Assay Details
Target Gene Details
Entrez Gene ID: | 328779 |
Gene Name: | heparan sulfate (glucosamine) 3-O-sulfotransferase 6 |
Gene Aliases: |
- |
Location: |
Chr.17:24752950-24758690 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hs3st6 | NM_001012402.1 | 2 | 858 | NP_001012402.1 |
| AK146974.1 | 3 | 997 | BAE27579.1 | |
| AY574375.1 | 2 | 858 | AAT84072.1 | |
| BC132520.1 | 2 | 845 | ||
| BC138814.1 | 2 | 845 | ||
| BC145424.1 | 2 | 815 |
More Information
Set Membership: |
|

Genomic Map