Assay Details
Target Gene Details
Entrez Gene ID: | 14370 |
Gene Name: | frizzled homolog 8 (Drosophila) |
Gene Aliases: |
Fz8 |
Location: |
Chr.18:9212856-9216201 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fzd8 | NM_008058.2 | 1 | 556 | NP_032084.2 |
| BC132543.1 | 1 | 556 | ||
| BC138062.1 | 1 | 556 |
Target Gene Details
Entrez Gene ID: | 225152 |
Gene Name: | gap junction protein, delta 4 |
Gene Aliases: |
9430022F06Rik, Cx39 |
Location: |
Chr.18:9137290-9289808 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gjd4 |
More Information
Set Membership: |
|

Genomic Map