Assay Details
Target Gene Details
Entrez Gene ID: | 14587 |
Gene Name: | glial cell line derived neurotrophic factor family receptor alpha 3 |
Gene Aliases: |
GFRalpha3, Y15110 |
Location: |
Chr.18:34689903-34720387 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gfra3 | NM_010280.3 | 4 | 682 | NP_034410.3 |
| AB008833.1 | 4 | 560 | BAA23562.1 | |
| AF020305.1 | 4 | 515 | AAB70931.1 | |
| AF036163.1 | 4 | 576 | AAC24468.1 | |
| AF041842.1 | 4 | 565 | AAC23558.1 | |
| AF051766.1 | 4 | 577 | AAC24354.1 | |
| AK046542.1 | 4 | 609 | BAC32778.1 | |
| BC066202.1 | 4 | 682 | AAH66202.1 | |
| Y15110.1 | 4 | 596 | CAA75384.1 |
More Information
Set Membership: |
|

Genomic Map