Assay Details
Target Gene Details
Entrez Gene ID: | 225152 |
Gene Name: | gap junction protein, delta 4 |
Gene Aliases: |
9430022F06Rik, Cx39 |
Location: |
Chr.18:9137290-9289808 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gjd4 | NM_153086.5 | 1 | 211 | NP_694726.2 |
| AJ414562.1 | CAC93844.1 | |||
| AK034663.1 | 1 | 212 | BAC28788.1 | |
| AK053016.1 | 1 | 163 | BAC35240.1 | |
| BC137984.1 | 1 | 188 |
More Information
Set Membership: |
|

Genomic Map