Assay Details
Target Gene Details
Entrez Gene ID: | 195359 |
Gene Name: | tripartite motif-containing 40 |
Gene Aliases: |
Gm319, Gm35, Gm865 |
Location: |
Chr.17:36881598-36896743 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trim40 | NM_001033235.3 | 2 | 858 | NP_001028407.1 |
| NM_001172168.1 | 2 | 858 | NP_001165639.1 | |
| AK136498.1 | 2 | 779 | BAE23012.1 | |
| BC147336.1 | 1 | 549 | ||
| BC147337.1 | 1 | 549 | ||
| BC172037.1 | 1 | 549 |
More Information
Set Membership: |
|

Genomic Map