Assay Details
Target Gene Details
Entrez Gene ID: | 121022 |
Gene Name: | mitochondrial ribosomal protein S6 |
Gene Aliases: |
AW046321 |
Location: |
Chr.16:92058336-92112227 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrps6 | NM_080456.1 | NP_536704.1 | ||
| AB049943.1 | BAB40996.1 | |||
| AK165590.1 | BAE38278.1 | |||
| AK166374.1 | BAE38738.1 | |||
| AK167304.1 | BAE39408.1 | |||
| AK167316.1 | BAE39418.1 | |||
| AY061856.1 | AAL35740.1 | |||
| BC027271.1 | AAH27271.1 |
Target Gene Details
Entrez Gene ID: | 53881 |
Gene Name: | solute carrier family 5 (inositol transporters), member 3 |
Gene Aliases: |
AA623876, BF642829, Smit1 |
Location: |
Chr.16:92058322-92087473 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc5a3 | NM_017391.3 | 2 | 1948 | NP_059087.2 |
| AK133628.1 | 1 | 3152 | BAE21756.1 | |
| AK144718.1 | 2 | 1949 | BAE26029.1 | |
| BB024285.2 | 1 | 393 | ||
| BC140982.1 | 1 | 1495 | ||
| BC140983.1 | 1 | 1495 |
More Information
Set Membership: |
|

Genomic Map