Assay Details
Target Gene Details
Entrez Gene ID: | 22661 |
Gene Name: | zinc finger protein 148 |
Gene Aliases: |
2210405J08Rik, AI480666, AW045217, BERF-1, BFCOL1, ZBP-89, Znf148 |
Location: |
Chr.16:33380775-33503903 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp148 | NM_011749.4 | 9 | 5350 | NP_035879.1 |
| AF316548.1 | 9 | 5289 | AAK63003.1 | |
| AK028512.1 | 1 | 123 |
More Information
Set Membership: |
|

Genomic Map