Assay Details
Target Gene Details
Entrez Gene ID: | 20975 |
Gene Name: | synaptojanin 2 |
Gene Aliases: |
AI481647, SJ2, mKIAA0348 |
Location: |
Chr.17:5941280-6045553 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Synj2 | NM_001113352.2 | 1 | 147 | NP_001106823.1 |
| NM_001113353.2 | 1 | 147 | NP_001106824.1 | |
| AF041857.1 | 1 | 148 | AAC40141.1 | |
| AK142919.1 | ||||
| BC058749.1 | AAH58749.1 |
More Information
Set Membership: |
|

Genomic Map