Assay Details
Target Gene Details
Entrez Gene ID: | 12193 |
Gene Name: | zinc finger protein 36, C3H type-like 2 |
Gene Aliases: |
Brf2, ERF2, Tis11d |
Location: |
Chr.17:84183924-84187947 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp36l2 | NM_001001806.2 | 1 | 207 | NP_001001806.1 |
| AK159513.1 | 1 | 208 | ||
| AK160955.1 | 1 | 206 | BAE36112.1 | |
| AK170763.1 | 1 | 208 | BAE42014.1 | |
| BB611978.1 | 1 | 206 | ||
| BB656287.1 | 1 | 208 | ||
| BC139416.1 | 1 | 83 |
More Information
Set Membership: |
|

Genomic Map