Assay Details
Target Gene Details
Entrez Gene ID: | 225164 |
Gene Name: | mindbomb homolog 1 (Drosophila) |
Gene Aliases: |
DIP-1, E430019M12Rik, Mib, mKIAA1323 |
Location: |
Chr.18:10725625-10818578 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mib1 | NM_144860.2 | 1 | 757 | NP_659109.2 |
| AK139338.1 | 1 | 638 | BAE23964.1 | |
| AY147848.1 | 1 | 256 | AAN18022.1 | |
| AY149907.1 | 1 | 77 | AAN75492.1 | |
| AY245539.1 | 1 | 757 | AAO91933.1 | |
| AY974091.1 | 1 | 50 | AAX84653.1 | |
| BC131662.1 | 2 | 223 |
More Information
Set Membership: |
|

Genomic Map