Assay Details
Target Gene Details
Entrez Gene ID: | 77128 |
Gene Name: | CREB3 regulatory factor |
Gene Aliases: |
A930001N09Rik, LRF |
Location: |
Chr.17:26714711-26780985 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Crebrf | NM_029870.2 | 4 | 797 | NP_084146.1 |
| AK020796.1 | 4 | 744 | BAB32214.1 | |
| AK030092.1 | 4 | 798 | BAC26779.1 | |
| AK044760.1 | 4 | 766 | BAC32070.1 | |
| AK132423.1 | 4 | 771 | ||
| AK132454.1 | 3 | 543 | ||
| AK137861.1 | 1 | 1216 | ||
| BC113191.1 | 4 | 691 | ||
| BC113192.1 | 4 | 691 |

Genomic Map