Assay Details
Target Gene Details
Entrez Gene ID: | 193796 |
Gene Name: | lysine (K)-specific demethylase 4B |
Gene Aliases: |
4732474L06Rik, Jmjd2b, mKIAA0876 |
Location: |
Chr.17:56326033-56403769 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kdm4b | NM_172132.2 | 12 | 1786 | NP_742144.1 |
| AK028962.1 | BAC26214.1 | |||
| AK078074.1 | 12 | 1779 | ||
| AK129233.1 | 12 | 1745 | BAC98043.1 | |
| AK141879.1 | 12 | 1763 | BAE24866.1 | |
| AK190105.1 | 2 | 338 | ||
| BC007145.1 | 12 | 1756 | AAH07145.1 |
More Information
Set Membership: |
|

Genomic Map