Assay Details
Target Gene Details
Entrez Gene ID: | 20975 |
Gene Name: | synaptojanin 2 |
Gene Aliases: |
AI481647, SJ2, mKIAA0348 |
Location: |
Chr.17:5941280-6045553 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Synj2 | NM_001113351.1 | 2 | 519 | NP_001106822.1 |
| NM_001113352.2 | 3 | 608 | NP_001106823.1 | |
| NM_001113353.2 | 3 | 608 | NP_001106824.1 | |
| NM_011523.2 | 2 | 519 | NP_035653.2 | |
| AF041857.1 | 3 | 609 | AAC40141.1 | |
| AF041862.1 | 2 | 464 | AAC40146.1 | |
| AK142919.1 | ||||
| AK148053.1 | 3 | 347 | ||
| AK163512.1 | ||||
| BC058749.1 | 3 | 485 | AAH58749.1 | |
| BC060214.1 | 2 | 365 | AAH60214.1 |
More Information
Set Membership: |
|

Genomic Map