Assay Details
Target Gene Details
Entrez Gene ID: | 321007 |
Gene Name: | serine active site containing 1 |
Gene Aliases: |
4930511N22Rik, D17Ertd141e |
Location: |
Chr.17:6040571-6079762 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serac1 | NM_001111017.1 | 17 | 2074 | NP_001104487.1 |
| NM_177311.4 | 16 | 1984 | NP_796285.2 | |
| AK019677.1 | 16 | 1973 | BAB31833.1 | |
| AK154605.1 | 17 | 2025 | BAE32707.1 | |
| AK155571.1 | 17 | 2077 | BAE33329.1 | |
| AK162246.1 | BAE36813.1 | |||
| AK170498.1 | 17 | 2025 | BAE41837.1 | |
| BC119594.1 | 17 | 2063 | ||
| BC119595.1 | 17 | 2063 |
Target Gene Details
Entrez Gene ID: | 20975 |
Gene Name: | synaptojanin 2 |
Gene Aliases: |
AI481647, SJ2, mKIAA0348 |
Location: |
Chr.17:5941280-6045553 on Build GRCm38 |
Assay Gene Location: | Within Exon 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Synj2 | NM_001113351.1 | 25 | 3829 | NP_001106822.1 |
| NM_011523.2 | 26 | 3964 | NP_035653.2 | |
| AF041860.1 | 12 | 1761 | AAC40144.1 | |
| AF041862.1 | 26 | 3909 | AAC40146.1 | |
| AK029755.1 | 23 | 3336 |
More Information
Set Membership: |
|

Genomic Map