Assay Details
Target Gene Details
Entrez Gene ID: | 224691 |
Gene Name: | zinc finger protein 472 |
Gene Aliases: |
KRIM-1, Krim-1A, Krim-1B, Krim1 |
Location: |
Chr.17:32965831-32979211 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp472 | NM_153063.3 | 4 | 627 | NP_694703.2 |
| AK142395.1 | 4 | 618 | BAE25054.1 | |
| AY195875.1 | 4 | 562 | AAP34297.1 | |
| BB652931.1 | ||||
| BC027407.1 | 4 | 617 | AAH27407.1 | |
| BQ885210.1 | 1 | 148 |

Genomic Map