Assay Details
Target Gene Details
Entrez Gene ID: | 77128 |
Gene Name: | CREB3 regulatory factor |
Gene Aliases: |
A930001N09Rik, LRF |
Location: |
Chr.17:26714711-26780985 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Crebrf | NM_029870.2 | 9 | 3181 | NP_084146.1 |
| AK030092.1 | 9 | 3182 | BAC26779.1 | |
| AK137518.1 | 1 | 310 | ||
| AK164174.1 | 1 | 589 |

Genomic Map