Assay Details
Target Gene Details
Entrez Gene ID: | 68636 |
Gene Name: | fumarylacetoacetate hydrolase domain containing 1 |
Gene Aliases: |
1110025H10Rik, AV002085 |
Location: |
Chr.17:24848896-24850302 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fahd1 | NM_023480.2 | 1 | 241 | NP_075969.1 |
| AB041600.1 | 1 | 75 | BAA95083.1 | |
| AK003914.1 | ||||
| AK044920.1 | BAC32142.1 | |||
| AK141973.1 | 1 | 241 | BAE24904.1 | |
| AK157292.1 | 1 | 241 | BAE34035.1 | |
| BC026949.1 | 1 | 227 | AAH26949.1 |
Target Gene Details
Entrez Gene ID: | 14651 |
Gene Name: | hydroxyacyl glutathione hydrolase |
Gene Aliases: |
BC019817, Glo-2, Glo2, Rsp29 |
Location: |
Chr.17:24840079-24864450 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hagh |
More Information
Set Membership: |
|

Genomic Map