Assay Details
Target Gene Details
Entrez Gene ID: | 106557 |
Gene Name: | lactate dehydrogenase A-like 6B |
Gene Aliases: |
4933402O15Rik, AI326310, LDHL, Ldhal |
Location: |
Chr.17:5417323-5418767 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ldhal6b | NM_175349.2 | 1 | 967 | NP_780558.1 |
| AK077089.1 | 1 | 913 | BAC36605.1 | |
| AY280363.1 | 1 | 967 | AAP35085.1 | |
| BC061193.1 | 1 | 912 | AAH61193.1 |
Target Gene Details
Entrez Gene ID: | 70544 |
Gene Name: | transmembrane protein 242 |
Gene Aliases: |
1110008A10Rik, 2310046K16Rik, 5730437N04Rik |
Location: |
Chr.17:5410864-5440260 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem242 | NM_027457.4 | NP_081733.3 | ||
| AK009851.1 | BAC25272.1 | |||
| AK043276.1 | ||||
| AK088025.1 | BAC40103.1 | |||
| AK155326.1 | BAE33193.1 | |||
| AK158107.1 | BAE34359.1 | |||
| BC019420.1 | AAH19420.1 |
More Information
Set Membership: |
|

Genomic Map