Assay Details
Target Gene Details
Entrez Gene ID: | 21354 |
Gene Name: | transporter 1, ATP-binding cassette, sub-family B (MDR/TAP) |
Gene Aliases: |
ABC17, APT1, Abcb2, Ham-1, Ham1, MTP1, PSF1, RING4, TAP, Tap-1, Y3 |
Location: |
Chr.17:34187556-34197225 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tap1 | NM_001161730.1 | 12 | 2527 | NP_001155202.1 |
| NM_013683.2 | 11 | 2611 | NP_038711.2 | |
| AK040919.1 | 9 | 3574 | ||
| AK151481.1 | 11 | 2366 | ||
| AK152953.1 | 11 | 2311 | BAE31620.1 | |
| AK153890.1 | 11 | 2615 | BAE32237.1 | |
| AK162731.1 | 11 | 2411 | BAE37041.1 | |
| AK166046.1 | 11 | 2614 | ||
| AK167485.1 | 11 | 2613 | ||
| AK171363.1 | 12 | 2282 | BAE42411.1 | |
| BC013802.1 | 11 | 2022 | AAH13802.1 | |
| BC024897.1 | 11 | 2134 | AAH24897.1 | |
| M55637.1 | 11 | 2003 | AAA39570.1 |
More Information
Set Membership: |
|

Genomic Map