Assay Details
Target Gene Details
Entrez Gene ID: | 21355 |
Gene Name: | transporter 2, ATP-binding cassette, sub-family B (MDR/TAP) |
Gene Aliases: |
ABC18, AI462429, APT2, Abcb3, Ham-2, Ham2, MTP2, PSF2, RING11, Tap-2, Y1, jas |
Location: |
Chr.17:34204085-34216321 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tap2 | NM_011530.3 | 12 | 2354 | NP_035660.3 |
| AK020022.1 | 11 | 2430 | ||
| AK079667.1 | 12 | 2268 | ||
| AK145234.1 | 12 | 2313 | ||
| AK151376.1 | 12 | 2264 | BAE30348.1 | |
| AK151483.1 | 12 | 2264 | BAE30436.1 | |
| AK151657.1 | 12 | 2287 | ||
| AK153287.1 | 12 | 2264 | BAE31870.1 | |
| AK159200.1 | 12 | 2320 | BAE34894.1 | |
| AK161091.1 | 12 | 2270 | ||
| AK169645.1 | 9 | 1562 | ||
| BC005578.1 | 12 | 2254 | AAH05578.1 | |
| BC051257.1 | 12 | 2267 | AAH51257.2 | |
| BE691532.1 | 1 | 137 | ||
| U60087.1 | AAB41970.1 |
More Information
Set Membership: |
|

Genomic Map