Assay Details
Target Gene Details
Entrez Gene ID: | 18440 |
Gene Name: | purinergic receptor P2X, ligand-gated ion channel, 6 |
Gene Aliases: |
P2rxl1, P2x6, P2xm |
Location: |
Chr.16:17561855-17572012 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| P2rx6 | NM_001159561.1 | 11 | 1724 | NP_001153033.1 |
| NM_011028.2 | 12 | 1805 | NP_035158.2 | |
| AK081308.1 | 12 | 1682 | ||
| AK138541.1 | 12 | 1657 | BAE23697.1 |
More Information
Set Membership: |
|

Genomic Map