Assay Details
Target Gene Details
Entrez Gene ID: | 12607 |
Gene Name: | CCAAT/enhancer binding protein zeta |
Gene Aliases: |
AI848081, CBF2, Cbf, Cebpa-rs1 |
Location: |
Chr.17:78919003-78937070 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cebpz | NM_001024806.2 | 16 | 3736 | NP_001019977.1 |
| AI848081.1 | ||||
| AK017537.1 | 2 | 1366 | ||
| AK159081.1 | 16 | 3733 | BAE34799.1 |
Target Gene Details
Entrez Gene ID: | 68554 |
Gene Name: | CCAAT/enhancer binding protein (C/EBP), zeta, opposite strand |
Gene Aliases: |
1110001A16Rik |
Location: |
Chr.17:78916499-78921048 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cebpzos | NM_001177402.1 | NP_001170873.1 | ||
| NM_001177403.1 | NP_001170874.1 | |||
| AK003199.1 | BAC25024.2 | |||
| AK027910.1 | ||||
| AV491230.1 | ||||
| BY102137.1 |
More Information
Set Membership: |
|

Genomic Map