Assay Details
Target Gene Details
Entrez Gene ID: | 19732 |
Gene Name: | ral guanine nucleotide dissociation stimulator-like 2 |
Gene Aliases: |
KE1.5, Rab2l, Rgt2, Rlf |
Location: |
Chr.17:33929435-33937687 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rgl2 | NM_009059.2 | 7 | 1086 | NP_033085.2 |
| AK144440.1 | 7 | 1088 | ||
| BC068121.1 | 7 | 1086 | AAH68121.1 | |
| KJ650210.1 | 6 | 894 | ||
| U54639.1 | 7 | 1015 | AAC52724.1 |
More Information
Set Membership: |
|

Genomic Map