Assay Details
Target Gene Details
Entrez Gene ID: | 72726 |
Gene Name: | tubulin-specific chaperone C |
Gene Aliases: |
2810055C19Rik |
Location: |
Chr.17:46890621-46892463 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbcc | NM_178385.3 | 1 | 1531 | NP_848472.2 |
More Information
Set Membership: |
|

Genomic Map