Assay Details
Target Gene Details
Entrez Gene ID: | 70573 |
Gene Name: | TBCC domain containing 1 |
Gene Aliases: |
5730478M09Rik, AU015083, BB165076 |
Location: |
Chr.16:22813215-22857701 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbccd1 | NM_001081368.1 | 4 | 825 | NP_001074837.1 |
| AK017705.1 | 4 | 832 | BAB30885.1 | |
| AK039905.1 | 4 | 650 | BAC30476.1 | |
| AK040787.1 | 4 | 797 | ||
| AK043059.1 | 4 | 799 | BAC31450.1 | |
| AK133804.1 | 4 | 766 | ||
| AK179171.1 | ||||
| BC082558.1 | 4 | 794 | AAH82558.1 |

Genomic Map