Assay Details
Target Gene Details
Entrez Gene ID: | 66467 |
Gene Name: | general transcription factor IIH, polypeptide 5 |
Gene Aliases: |
2700017P07Rik, 2810432H05Rik, D17Wsu155e |
Location: |
Chr.17:6079828-6085488 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gtf2h5 | NM_181392.3 | 3 | 1002 | NP_852057.2 |
| AK013225.1 | 3 | 966 | BAC25398.1 | |
| BM940130.1 | 1 | 81 |
More Information
Set Membership: |
|

Genomic Map