Assay Details
Target Gene Details
Entrez Gene ID: | 574402 |
Gene Name: | G protein-coupled receptor 17 |
Gene Aliases: |
AI853548 |
Location: |
Chr.18:31942999-31949636 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpr17 | NM_001025381.2 | 2 | 2683 | NP_001020552.1 |
| BC070439.1 | 2 | 2683 | AAH70439.1 |
Target Gene Details
Entrez Gene ID: | 225341 |
Gene Name: | LIM and senescent cell antigen like domains 2 |
Gene Aliases: |
PINCH2 |
Location: |
Chr.18:31922277-31958620 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lims2 | NM_144862.3 | NP_659111.1 | ||
| BC010816.1 | AAH10816.1 |
More Information
Set Membership: |
|

Genomic Map