Assay Details
Target Gene Details
Entrez Gene ID: | 23984 |
Gene Name: | phosphodiesterase 10A |
Gene Aliases: |
- |
Location: |
Chr.17:8525187-8986648 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pde10a | NM_001290707.1 | 23 | 6054 | NP_001277636.1 |
| NM_011866.2 | 22 | 6008 | NP_035996.2 | |
| AK082893.1 | 1 | 1584 | ||
| AK147690.1 | 22 | 5926 | ||
| AY360383.1 | 22 | 6008 | AAR12579.1 |
More Information
Set Membership: |
|

Genomic Map