Assay Details
Target Gene Details
Entrez Gene ID: | 225865 |
Gene Name: | cation channel, sperm associated 1 |
Gene Aliases: |
Catsper, KSper |
Location: |
Chr.19:5335602-5344281 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Catsper1 | NM_139301.2 | 1 | 770 | NP_647462.1 |
| AF407332.1 | 1 | 770 | AAL14104.1 |
More Information
Set Membership: |
|

Genomic Map