Assay Details
Target Gene Details
Entrez Gene ID: | 70445 |
Gene Name: | CD248 antigen, endosialin |
Gene Aliases: |
2610111G01Rik, AI842296, Cd164l1, Tem1 |
Location: |
Chr.19:5068078-5070640 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd248 | NM_054042.2 | 1 | 423 | NP_473383.1 |
| AF378758.1 | 1 | 381 | AAL11995.1 | |
| AF388572.1 | 1 | 378 | AAK84664.1 | |
| AK144655.1 | 1 | 393 | BAE25992.1 | |
| BC046318.1 | 1 | 423 | AAH46318.1 |

Genomic Map