Assay Details
Target Gene Details
Entrez Gene ID: | 54632 |
Gene Name: | FtsJ homolog 1 (E. coli) |
Gene Aliases: |
AI931847, Ftsj, Ftsjl, Sfc12 |
Location: |
Chr.X:8238668-8252406 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ftsj1 | NM_001290430.1 | 13 | 3063 | NP_001277359.1 |
| NM_133991.2 | 13 | 3069 | NP_598752.2 | |
| AK036313.1 | 13 | 3027 | BAC29380.1 | |
| BU610453.1 | 1 | 426 |
More Information
Set Membership: |
|

Genomic Map