Assay Details
Target Gene Details
Entrez Gene ID: | 12686 |
Gene Name: | elongation of very long chain fatty acids (FEN1/Elo2, SUR4/Elo3, yeast)-like 3 |
Gene Aliases: |
CIN-2, Cig30 |
Location: |
Chr.19:46131899-46135694 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Elovl3 | NM_007703.2 | 1 | 90 | NP_031729.1 |
| BC016468.1 | 1 | 85 | AAH16468.1 | |
| U97107.1 | 1 | 90 | AAC06127.1 |
More Information
Set Membership: |
|

Genomic Map