Assay Details
Target Gene Details
Entrez Gene ID: | 60367 |
Gene Name: | interleukin 1 receptor accessory protein-like 2 |
Gene Aliases: |
- |
Location: |
Chr.X:137571549-138846955 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il1rapl2 | NM_030688.1 | NP_109613.1 | ||
| AF284437.1 | AAG21371.1 | |||
| BC119274.1 | ||||
| BC120555.1 |
Target Gene Details
Entrez Gene ID: | 67944 |
Gene Name: | testis expressed 13A |
Gene Aliases: |
1700025D03Rik |
Location: |
Chr.X:138208158-138209591 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tex13a | NM_026469.2 | 1 | 406 | NP_080745.2 |
| AK006452.1 | 1 | 406 |
More Information
Set Membership: |
|

Genomic Map