Assay Details
Target Gene Details
Entrez Gene ID: | 240255 |
Gene Name: | YTH domain containing 2 |
Gene Aliases: |
3010002F02Rik, BC037178 |
Location: |
Chr.18:44827744-44889720 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ythdc2 | NM_001163013.1 | 1 | 210 | NP_001156485.1 |
| BC138263.1 | 1 | 210 | ||
| BC171951.1 | 1 | 210 |
More Information
Set Membership: |
|

Genomic Map