Assay Details
Target Gene Details
Entrez Gene ID: | 12686 |
Gene Name: | elongation of very long chain fatty acids (FEN1/Elo2, SUR4/Elo3, yeast)-like 3 |
Gene Aliases: |
CIN-2, Cig30 |
Location: |
Chr.19:46131899-46135694 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Elovl3 | NM_007703.2 | 4 | 1659 | NP_031729.1 |
| AA241332.1 | 1 | 268 | ||
| U97107.1 | 4 | 1659 | AAC06127.1 |
Target Gene Details
Entrez Gene ID: | 18742 |
Gene Name: | paired-like homeodomain transcription factor 3 |
Gene Aliases: |
Ptx3, ak |
Location: |
Chr.19:46135294-46148325 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pitx3 |
More Information
Set Membership: |
|

Genomic Map