Assay Details
Target Gene Details
Entrez Gene ID: | 102902673 |
Gene Name: | predicted gene 21992 |
Gene Aliases: |
Rbm14-rbm4 |
Location: |
Chr.19:4784293-4811634 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm21992 | NM_001290127.1 | 4 | 2422 | NP_001277056.1 |
| NM_001290128.1 | 3 | 1999 | NP_001277057.1 |
Target Gene Details
Entrez Gene ID: | 19653 |
Gene Name: | RNA binding motif protein 4 |
Gene Aliases: |
4921506I22Rik, Lark1, Mlark, Rbm4a, lark |
Location: |
Chr.19:4784293-4794009 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rbm4 | NM_001290122.1 | 4 | 2153 | NP_001277051.1 |
| NM_001290123.1 | 4 | 2142 | NP_001277052.1 | |
| NM_001290124.1 | 3 | 1471 | NP_001277053.1 | |
| NM_001290125.1 | 3 | 1507 | NP_001277054.1 | |
| NM_009032.3 | 4 | 2189 | NP_033058.2 | |
| AK049659.1 | 4 | 2150 | BAC33862.1 | |
| BM932360.1 |
More Information
Set Membership: |
|

Genomic Map