Assay Details
Target Gene Details
Entrez Gene ID: | 226016 |
Gene Name: | abhydrolase domain containing 17B |
Gene Aliases: |
5730446C15Rik, Cgi67, Fam108b, Fam108b1 |
Location: |
Chr.19:21653309-21685638 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abhd17b | NM_146096.3 | 1 | 51 | NP_666208.2 |
| AK017641.1 | 1 | 46 | ||
| AK050879.1 | 1 | 176 |
More Information
Set Membership: |
|

Genomic Map