Assay Details
Target Gene Details
Entrez Gene ID: | 93761 |
Gene Name: | SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 1 |
Gene Aliases: |
5730494M04Rik, Snf2l |
Location: |
Chr.X:47809370-47892931 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smarca1 | NM_001290708.1 | 1 | 233 | NP_001277637.1 |
| NM_053123.4 | 1 | 233 | NP_444353.3 | |
| AF325920.1 | 1 | 237 | AAK52453.1 | |
| AK030741.1 | 1 | 200 | BAC27109.1 | |
| AK032978.1 | 1 | 201 | ||
| AK035068.1 | 1 | 113 | BAC28931.1 | |
| AK038470.1 | 2 | 152 | ||
| AK052092.1 | 1 | 204 | ||
| BC016071.1 | 1 | 210 | ||
| BC057115.1 | 1 | 233 | AAH57115.1 | |
| BU704042.1 | 1 | 233 |
More Information
Set Membership: |
|

Genomic Map