Assay Details
Target Gene Details
Entrez Gene ID: | 68034 |
Gene Name: | family with sequence similarity 122, member A |
Gene Aliases: |
2410124L17Rik, 2900009I07Rik, ENSMUSG00000051082, Gm9849 |
Location: |
Chr.19:24475779-24477474 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam122a | NM_026520.3 | 1 | 569 | NP_080796.1 |
| AK005219.1 | 1 | 562 | BAB23889.1 | |
| AK010777.1 | 1 | 581 | ||
| AK011741.1 | 1 | 553 | BAB27811.1 | |
| AK018219.1 | 1 | 524 | ||
| AK033646.1 | 1 | 569 | BAC28405.1 | |
| AK077654.1 | 1 | 551 | BAC36930.1 | |
| AK172089.1 | 1 | 554 | BAE42820.1 | |
| BC028637.1 | 1 | 537 | AAH28637.1 | |
| BC083062.1 | 1 | 559 | AAH83062.1 |
Target Gene Details
Entrez Gene ID: | 18719 |
Gene Name: | phosphatidylinositol-4-phosphate 5-kinase, type 1 beta |
Gene Aliases: |
PI4P5K-I[b], PIP5K1-beta, PIP5KIbeta, Pipk5b, STM7 |
Location: |
Chr.19:24294794-24555827 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pip5k1b | NM_008846.2 | NP_032872.1 | ||
| AK077334.1 | ||||
| AK133809.1 | ||||
| AK146100.1 | BAE26901.1 | |||
| AK149354.1 | BAE28830.1 | |||
| AK164552.1 | BAE37836.1 | |||
| BC034864.1 | AAH34864.1 |
More Information
Set Membership: |
|

Genomic Map