Assay Details
Target Gene Details
Entrez Gene ID: | 20689 |
Gene Name: | sal-like 3 (Drosophila) |
Gene Aliases: |
B130022O04Rik, Msal, Msal-1, Sal, Salt, Spalt |
Location: |
Chr.18:80966359-80986578 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sall3 | NM_178280.3 | 2 | 2536 | NP_840064.2 |
| AK045051.1 | 1 | 422 | BAC32197.1 | |
| BC072631.1 | 2 | 2480 | AAH72631.1 | |
| X97581.1 | 1 | 2408 | CAA66196.1 |
More Information
Set Membership: |
|

Genomic Map