Assay Details
Target Gene Details
Entrez Gene ID: | 16814 |
Gene Name: | ladybird homeobox homolog 1 (Drosophila) |
Gene Aliases: |
Lbx1h |
Location: |
Chr.19:45232684-45237435 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lbx1 | NM_010691.5 | 1 | 109 | NP_034821.2 |
| AB108497.1 | 1 | 109 | BAC75634.1 | |
| AK144562.1 | BAE25939.1 | |||
| BC119177.1 | 1 | 198 | ||
| BC120586.1 | 1 | 198 | AAI20587.1 | |
| X90829.1 | 1 | 609 | CAA62343.1 |
More Information
Set Membership: |
|

Genomic Map