Assay Details
Target Gene Details
Entrez Gene ID: | 68642 |
Gene Name: | transmembrane protein 216 |
Gene Aliases: |
1110017C22Rik, 2810441K11Rik, 4921533J23Rik, A930021F15Rik, AI482550 |
Location: |
Chr.19:10539326-10556303 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem216 | NM_001277860.1 | 5 | 735 | NP_001264789.1 |
| NM_001277861.1 | 5 | 749 | NP_001264790.1 | |
| NM_026798.3 | 5 | 808 | NP_081074.1 | |
| AF332866.1 | 4 | 674 | AAK17992.1 | |
| AK003740.1 | 5 | 810 | BAB22973.1 | |
| AK013286.1 | 5 | 739 | ||
| BC052053.1 | 5 | 712 | AAH52053.1 | |
| BX633507.1 | 1 | 296 | ||
| BY704000.1 | 5 | 810 | ||
| DV063021.1 | 5 | 803 |

Genomic Map