Assay Details
Target Gene Details
Entrez Gene ID: | 15108 |
Gene Name: | hydroxysteroid (17-beta) dehydrogenase 10 |
Gene Aliases: |
17bHSD10, Ads9, ERAB, Hadh2 |
Location: |
Chr.X:152001896-152004442 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hsd17b10 | NM_016763.2 | 6 | 760 | NP_058043.3 |
| AF233685.1 | 6 | 760 | AAK15008.1 | |
| AK002368.1 | 6 | 755 | ||
| AK013340.1 | 6 | 756 | BAB28800.1 | |
| AK077735.1 | 6 | 754 | BAC36987.1 | |
| AK088685.1 | 6 | 776 | BAC40505.1 | |
| AK179857.1 | ||||
| BC027517.1 | 6 | 760 | AAH27517.1 | |
| U96116.1 | 6 | 861 | AAB57689.1 |
More Information
Set Membership: |
|

Genomic Map