Assay Details
Target Gene Details
Entrez Gene ID: | 104174 |
Gene Name: | glycine decarboxylase |
Gene Aliases: |
D030049L12Rik, D19Wsu57e |
Location: |
Chr.19:30098441-30175441 on Build GRCm38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gldc | NM_138595.2 | 25 | 3450 | NP_613061.1 |
| AK010327.1 | 5 | 761 | BAB26854.1 | |
| AK080789.1 | 25 | 3422 | BAC38022.1 | |
| AK080937.1 | 25 | 3440 | ||
| BC017135.1 | 25 | 3427 | AAH17135.1 | |
| BF020198.1 | 1 | 343 |
More Information
Set Membership: |
|

Genomic Map