Assay Details
Target Gene Details
Entrez Gene ID: | 78891 |
Gene Name: | SCY1-like 1 (S. cerevisiae) |
Gene Aliases: |
2810011O19Rik, C85140, Ntkl, mdf, mfd, p105 |
Location: |
Chr.19:5758427-5771414 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Scyl1 | NM_023912.2 | 17 | 2463 | NP_076401.1 |
| AF276514.1 | 17 | 2463 | AAG17393.1 | |
| AK019240.1 | 16 | 2535 | ||
| AK144134.1 | 17 | 2449 | BAE25720.1 | |
| AK145869.1 | 17 | 2483 | BAE26711.1 | |
| BC034519.1 | 17 | 2449 | AAH34519.1 | |
| HM856325.1 | 17 | 2412 | ||
| HM856326.1 | 16 | 2542 |
More Information
Set Membership: |
|

Genomic Map