Assay Details
Target Gene Details
Entrez Gene ID: | 109113 |
Gene Name: | ubiquitin-like, containing PHD and RING finger domains 2 |
Gene Aliases: |
2310065A22Rik, AI426270, AW214556, D130071B19Rik, Nirf |
Location: |
Chr.19:30030433-30093729 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Uhrf2 | NM_144873.2 | 1 | 74 | NP_659122.2 |
| AF274047.1 | AAM33798.1 | |||
| AK031036.1 | 1 | 136 | BAC27224.1 | |
| AK041564.1 | 1 | 96 | BAC30987.1 | |
| AK042321.1 | 1 | 115 | BAC31223.1 | |
| AK051743.1 | 1 | 115 | BAC34750.1 | |
| AK080925.1 | 1 | 115 | BAC38081.1 | |
| AK082518.1 | 1 | 144 | ||
| AK172682.1 | BAE43129.1 | |||
| BC060241.1 | 1 | 74 | AAH60241.1 |
More Information
Set Membership: |
|

Genomic Map