Assay Details
Target Gene Details
Entrez Gene ID: | 78242 |
Gene Name: | serine peptidase inhibitor, Kazal type 12 |
Gene Aliases: |
9230117E20Rik |
Location: |
Chr.18:44104523-44108543 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spink12 | NM_030061.3 | 4 | 477 | NP_084337.1 |
| AK020352.1 | 4 | 378 | BAB32081.2 | |
| AK136616.1 | 4 | 478 | BAE23072.1 | |
| BC145914.1 | ||||
| BC145916.1 | ||||
| DQ437332.1 | 4 | 536 |
More Information
Set Membership: |
|

Genomic Map