Assay Details
Target Gene Details
Entrez Gene ID: | 23963 |
Gene Name: | teneurin transmembrane protein 1 |
Gene Aliases: |
Odz1, Odz2, TCAP-1, Ten-m1, ten-1 |
Location: |
Chr.X:42527866-43429156 on Build GRCm38 |
Assay Gene Location: | Within Exon 31 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tenm1 | NM_011855.4 | 31 | 6581 | NP_035985.2 |
| AB025410.1 | 31 | 6583 | BAA77396.1 | |
| AK037897.1 | 4 | 1270 | BAC29893.1 | |
| BC138860.1 | 30 | 6564 | ||
| BC138861.1 | 30 | 6564 |

Genomic Map