Assay Details
Target Gene Details
Entrez Gene ID: | 23825 |
Gene Name: | barrier to autointegration factor 1 |
Gene Aliases: |
Baf, Bcrp1, L2bp1/Baf |
Location: |
Chr.19:5364633-5366876 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Banf1 | NM_001038231.2 | 3 | 418 | NP_001033320.1 |
| NM_001286608.1 | 3 | 760 | NP_001273537.1 | |
| NM_011793.3 | 3 | 626 | NP_035923.1 | |
| NR_104482.1 | 2 | 487 | ||
| AB025349.1 | 3 | 358 | BAA83102.1 | |
| AF037080.1 | AAC40032.1 | |||
| AK003025.1 | 3 | 402 | BAB22518.1 | |
| AK004212.1 | 3 | 385 | BAB23223.1 | |
| AK010257.1 | 3 | 404 | BAB26800.1 | |
| AK011079.1 | 3 | 407 | BAB27383.1 | |
| AK011100.1 | 3 | 405 | BAB27397.1 | |
| AK012588.1 | 3 | 388 | BAB28337.1 | |
| AK088896.1 | 3 | 382 | BAC40639.1 | |
| AK168039.1 | 3 | 408 | ||
| AK185753.1 | 1 | 61 | ||
| BC003709.1 | 3 | 395 | AAH03709.1 | |
| CK780850.1 | 3 | 390 |

Genomic Map