Assay Details
Target Gene Details
Entrez Gene ID: | 14675 |
Gene Name: | guanine nucleotide binding protein, alpha 14 |
Gene Aliases: |
AU023208 |
Location: |
Chr.19:16435667-16610818 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gna14 | NM_008137.4 | 7 | 2150 | NP_032163.3 |
| AK035320.1 | 6 | 990 | BAC29032.1 | |
| AK085444.1 | 7 | 2056 | BAC39447.1 | |
| AK160808.1 | 7 | 1744 | BAE36026.1 | |
| AK168498.1 | 7 | 1806 | BAE40384.1 | |
| BC027015.1 | 7 | 1618 | AAH27015.1 | |
| M80631.1 | 7 | 1279 | AAA83222.1 |
More Information
Set Membership: |
|

Genomic Map